Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022924.2 | NP_075213.2 | 4 | 312 | ||
GenBank mRNA | EX492416.1 | - | 4 | 333 | ||
X52835.1 | - | 4 | 377 |