Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr1i3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270838.1 | NP_001257767.1 | 9 | 1176 | ||
NM_001270839.1 | NP_001257768.1 | 9 | 1154 | |||
NM_001270840.1 | NP_001257769.1 | 9 | 1173 | |||
NM_022941.4 | NP_075230.1 | 9 | 1158 | |||
XM_006250261.2 | XP_006250323.1 | 9 | 1264 | |||
XM_008769759.1 | XP_008767981.1 | 8 | 2121 | |||
GenBank mRNA | AB104736.1 | - | 9 | 1211 | ||
AB105072.1 | - | 8 | 1302 | |||
AB204897.1 | - | 8 | 1024 | |||
AB204898.1 | - | 8 | 1027 | |||
AB204899.1 | - | 8 | 1005 | |||
AB204900.1 | - | 7 | 1204 | |||
AF133094.1 | - | 8 | 1009 | |||
AF133095.1 | - | 8 | 1009 | |||
BC089763.1 | - | 9 | 1173 | 7324318 |