Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270701.1 | NP_001257630.1 | 7 | 1224 | ||
NM_001270702.1 | NP_001257631.1 | 7 | 1137 | |||
NM_023021.2 | NP_075410.2 | 7 | 1224 | |||
XM_006228466.2 | XP_006228528.1 | 7 | 2984 | |||
XM_006228467.2 | XP_006228529.1 | 7 | 2985 | |||
XM_006228469.1 | XP_006228531.1 | 8 | 1206 | |||
XM_006228470.2 | XP_006228532.1 | 7 | 1054 | |||
XM_008758975.1 | XP_008757197.1 | 8 | 2879 | |||
XM_008758976.1 | XP_008757198.1 | 7 | 1403 | |||
XM_008758977.1 | XP_008757199.1 | 6 | 3385 | |||
XM_008758978.1 | XP_008757200.1 | 7 | 1043 | |||
GenBank mRNA | AB292802.1 | - | 5 | 699 | ||
AF149250.1 | - | 7 | 1082 | |||
AF156554.1 | - | 7 | 1224 | |||
AF190458.1 | - | 7 | 1194 | |||
AJ133438.2 | - | 7 | 1082 | |||
BC128736.1 | - | 7 | 1175 | 8372655 | ||
EU872449.1 | - | 7 | 1197 | |||
EU872450.1 | - | 7 | 1197 | |||
EU872451.1 | - | 7 | 1110 |