Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024151.1 | NP_077065.1 | 6 | 696 | ||
GenBank mRNA | BC063167.1 | - | 6 | 654 | 6921397 | |
FQ213125.1 | - | 6 | 679 | |||
FQ213804.1 | - | 6 | 693 | |||
FQ234083.1 | - | 6 | 654 | |||
L12383.1 | - | 6 | 696 |