Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024151.1 | NP_077065.1 | 6 | 789 | ||
GenBank mRNA | BC063167.1 | - | 6 | 747 | 6921397 | |
FQ213125.1 | - | 6 | 772 | |||
FQ213804.1 | - | 6 | 787 | |||
L12383.1 | - | 6 | 789 | |||
M86705.1 | - | 1 | 30 |