Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024152.2 | NP_077066.1 | 1 | 535 | ||
GenBank mRNA | BC091146.1 | - | 1 | 535 | 7309902 | |
L12385.1 | - | 1 | 524 |