Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024155.3 | NP_077069.3 | 5 | 320 | ||
XM_006236841.2 | XP_006236903.1 | 6 | 407 | |||
XM_006236842.2 | XP_006236904.1 | 5 | 943 | |||
GenBank mRNA | BC085688.1 | - | 5 | 320 | 7191458 | |
D38224.2 | - | 3 | 229 |