Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gucy2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024380.1 | NP_077356.1 | 7 | 1657 | ||
GenBank mRNA | L36029.1 | - | 7 | 1657 |