Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrpd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082539.1 | NP_001076008.1 | 3 | 767 | ||
NM_001082540.1 | NP_001076009.1 | 4 | 824 | |||
NM_001082541.1 | NP_001076010.1 | 3 | 767 | |||
NM_024404.2 | NP_077380.2 | 4 | 824 | |||
GenBank mRNA | AB046615.1 | - | 4 | 588 | ||
AB046616.1 | - | 3 | 482 | |||
AB046617.1 | - | 4 | 539 | |||
AB046618.1 | - | 3 | 482 | |||
FQ212501.1 | - | 1 | 77 |
Gene Symbol: Hnrnpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | X16933.1 | - | 101 |