Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030846.2 | NP_110473.2 | 3, 4 | 364 | ||
GenBank mRNA | AA926364.1 | - | 3, 4 | 364 | ||
BC091144.1 | - | 2, 3 | 308 | 7382476 | ||
D49846.1 | - | 3, 4 | 479 | |||
X62853.1 | - | 2, 3 | 214 |