Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030853.1 | NP_110480.1 | 1, 2 | 346 | ||
XM_008759919.1 | XP_008758141.1 | 1, 2 | 800 | |||
GenBank mRNA | AJ001184.1 | - | 1, 2 | 346 | ||
BC088130.1 | - | 1, 2 | 306 | 7382238 | ||
FQ233731.1 | - | 1, 2 | 363 |