Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278282.1 | NP_001265211.1 | 7 | 1494 | ||
NM_001278283.1 | NP_001265212.1 | 8 | 1574 | |||
NM_001278285.1 | NP_001265214.1 | 5 | 1402 | |||
NM_001278286.1 | NP_001265215.1 | 6 | 1418 | |||
NM_001278288.1 | NP_001265217.1 | 5 | 1409 | |||
NM_001278289.1 | NP_001265218.1 | 7 | 1558 | |||
NM_001318926.1 | NP_001305855.1 | 7 | 1543 | |||
NM_001318927.1 | NP_001305856.1 | 7 | 1587 | |||
NM_001318928.1 | NP_001305857.1 | 7 | 1438 | |||
NM_001318929.1 | NP_001305858.1 | 6 | 1485 | |||
NM_001318930.1 | NP_001305859.1 | 6 | 1478 | |||
NM_001318931.1 | NP_001305860.1 | 6 | 1440 | |||
NM_006916.2 | NP_008847.1 | 6 | 1463 | |||
NM_199229.2 | NP_954699.1 | 6 | 1489 | |||
XM_006712677.3 | XP_006712740.1 | 7 | 1436 | |||
GenBank mRNA | AB075523.1 | - | 1 | 114 | ||
AJ224326.1 | - | 5 | 1231 | |||
AK056028.1 | - | 6 | 1400 | |||
AK093658.1 | - | 8 | 1505 | |||
AK096199.1 | - | 7 | 1325 | |||
AK303184.1 | - | 5 | 1329 | |||
AK308741.1 | - | 6 | 1327 | |||
BC005148.2 | - | 6 | 1397 | 3505010 | ||
BC016764.1 | - | 6 | 1426 | 4107315 | ||
BC072401.1 | - | 6 | 1398 | 6047438 |