Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030863.1 | NP_110490.1 | 3, 4 | 298 | ||
GenBank mRNA | AF004811.1 | - | 3, 4 | 298 |