Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031028.3 | NP_112290.2 | 6 | 844 | ||
XM_006255879.2 | XP_006255941.1 | 6 | 938 | |||
XM_008772700.1 | XP_008770922.1 | 5 | 917 | |||
GenBank mRNA | AB016160.1 | - | 1 | 387 | ||
AB016161.1 | - | 1 | 387 | |||
AF283276.1 | - | 5 | 758 | |||
AJ831471.1 | - | 6 | 842 | |||
AJ831472.1 | - | 6 | 966 | |||
Y10369.2 | - | 6 | 816 | |||
Y10370.2 | - | 1 | 489 |
Set Membership: |
G-protein coupled receptor |