Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270621.1 | NP_001257550.1 | 11 | 1631 | ||
NM_031075.2 | NP_112337.2 | 12 | 1703 | |||
GenBank mRNA | BC081783.1 | - | 13 | 1725 | 7134300 | |
X90651.1 | - | 12 | 1703 | |||
X91167.1 | - | 12 | 1693 |