Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004132.1 | NP_001004132.1 | 5, 6 | 540 | ||
NM_031077.1 | NP_112339.1 | 5, 6 | 899 | |||
XM_006256617.2 | XP_006256679.1 | 5, 6 | 4667 | |||
XM_006256618.2 | XP_006256680.1 | 5, 6 | 900 | |||
XM_006256619.2 | XP_006256681.1 | 5, 6 | 952 | |||
XM_008773037.1 | XP_008771259.1 | 5, 6 | 4670 | |||
GenBank mRNA | BC078711.1 | - | 5, 6 | 540 | 7112874 | |
U36444.1 | - | 5, 6 | 899 |