Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031090.2 | NP_112352.2 | 6 | 661 | ||
GenBank mRNA | AY321327.1 | - | 617 | |||
BC066662.1 | - | 6 | 661 | 5623442 | ||
FQ209441.1 | - | 6 | 672 | |||
FQ213656.1 | - | 6 | 671 | |||
FQ214472.1 | - | 6 | 671 |