Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006944.2 | NP_008875.1 | 3 | 371 | ||
XM_005246102.3 | XP_005246159.1 | 3 | 402 | |||
XM_011511698.2 | XP_011510000.1 | 3 | 402 | |||
XM_011511699.2 | XP_011510001.1 | 3 | 402 | |||
XM_011511700.2 | XP_011510002.1 | 3 | 402 | |||
GenBank mRNA | AJ308099.1 | - | 3 | 371 | ||
BC069401.1 | - | 3 | 286 | 7262158 | ||
BC106705.1 | - | 3 | 286 | |||
BC112438.1 | - | 3 | 286 | |||
BC113972.1 | - | 2 | 161 | |||
U20530.1 | - | 3 | 286 |