Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Soat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031118.1 | NP_112380.1 | 6 | 492 | ||
XM_008769681.1 | XP_008767903.1 | 5 | 741 | |||
GenBank mRNA | D86373.1 | - | 6 | 492 |