Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031125.1 | NP_112387.1 | 4, 5 | 339 | ||
XM_006230345.2 | XP_006230407.1 | 4, 5 | 846 | |||
XM_006230346.2 | XP_006230408.1 | 4, 5 | 871 | |||
XM_006230348.2 | XP_006230410.1 | 4, 5 | 836 | |||
GenBank mRNA | L20821.1 | - | 4, 5 | 339 |