Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006948.4 | NP_008879.3 | 5 | 2135 | ||
GenBank mRNA | AK128540.1 | - | 5 | 1917 | ||
AK223282.1 | - | 5 | 2093 | |||
BC036370.2 | - | 5 | 2097 | 4837136 | ||
CA392315.1 | - | 1 | 75 | |||
U04735.1 | - | 5 | 2101 |