Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031140.1 | NP_112402.1 | 1 | 174 | ||
GenBank mRNA | BC061847.1 | - | 1 | 214 | 5621065 | |
X62952.1 | - | 1 | 174 |