Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031146.2 | NP_112408.1 | 3 | 285 | ||
GenBank mRNA | AF315378.1 | - | 2 | 170 | ||
BC059131.1 | - | 3 | 265 | 6921520 | ||
DY310102.1 | - | 3 | 285 | |||
FQ214345.1 | - | 3 | 259 |