Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc20a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031148.1 | NP_112410.1 | 1 | 251 | ||
XM_006234991.2 | XP_006235053.1 | 1 | 475 | |||
XM_006234992.2 | XP_006235054.1 | 1 | 475 | |||
GenBank mRNA | AB000489.1 | - | 1 | 291 |