Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPK1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006952.3 | NP_008883.2 | 8 | 1184 | ||
GenBank mRNA | AB002155.1 | - | 8 | 1142 | ||
AK225868.1 | - | 7 | 1075 | |||
BC063568.1 | - | 7 | 1132 | 3861427 |