Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dynlt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031318.1 | NP_112608.1 | 3 | 132 | ||
XM_006227901.2 | XP_006227963.1 | 3 | 197 | |||
GenBank mRNA | AB010119.1 | - | 3 | 121 | ||
AJ131437.1 | - | 3 | 132 | |||
BC166879.1 | - | 3 | 164 | 6919674 |