Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF33B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305033.1 | NP_001291962.1 | 4 | 6089 | ||
NM_001305035.1 | NP_001291964.1 | 6 | 6219 | |||
NM_001305036.1 | NP_001291965.1 | 5 | 6147 | |||
NM_001305037.1 | NP_001291966.1 | 5 | 6074 | |||
NM_001305038.1 | NP_001291967.1 | 5 | 6166 | |||
NM_001305039.1 | NP_001291968.1 | 6 | 6224 | |||
NM_001305040.1 | NP_001291969.1 | 7 | 6300 | |||
NM_006955.2 | NP_008886.1 | 5 | 6142 | |||
GenBank mRNA | AJ491697.1 | - | 5 | 5849 |