Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031326.1 | NP_112616.1 | 4 | 411 | ||
GenBank mRNA | AB014089.1 | - | 4 | 411 | ||
AF246196.1 | - | 3 | 188 | |||
AF377866.1 | - | 4 | 418 | |||
AJ312746.1 | - | 4 | 411 | |||
BC062022.1 | - | 4 | 413 | 5623491 |