Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timeless | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031340.1 | NP_112630.1 | 29 | 4148 | ||
GenBank mRNA | AB019576.1 | - | 29 | 4148 |