Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031341.1 | NP_112631.1 | 2 | 296 | ||
XM_006252020.2 | XP_006252082.1 | 2 | 232 | |||
XM_006252021.2 | XP_006252083.1 | 5 | 701 | |||
XM_006252023.2 | XP_006252085.1 | 2 | 237 | |||
XM_006252024.2 | XP_006252086.1 | 2 | 926 | |||
XM_008770705.1 | XP_008768927.1 | 4 | 644 | |||
GenBank mRNA | AB020520.1 | - | 2 | 296 | ||
AF200684.1 | - | 1 | 104 | |||
BC091142.1 | - | 2 | 211 | 7129657 |