Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vdac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031354.1 | NP_112644.1 | 6, 7 | 752 | ||
XM_006251661.1 | XP_006251723.1 | 7, 8 | 873 | |||
GenBank mRNA | AB039663.1 | - | 6, 7 | 752 | ||
AF268468.1 | - | 5, 6 | 588 | |||
BC063164.1 | - | 6, 7 | 721 | 6921675 | ||
FQ217146.1 | - | 3, 4 | 477 | |||
FQ219952.1 | - | 3, 4 | 477 | |||
FQ228796.1 | - | 3, 4 | 477 |
Gene Symbol: LOC362126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761811.1 | XP_008760033.1 | 1 | 581 |