Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031360.2 | NP_112650.1 | 3, 4 | 493 | ||
XM_008773007.1 | XP_008771229.1 | 4, 5 | 430 | |||
GenBank mRNA | AB047002.1 | - | 3, 4 | 232 | ||
BC091139.1 | - | 3, 4 | 493 | 7378100 |