Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031507.1 | NP_113695.1 | 18 | 2278 | ||
XM_008770416.1 | XP_008768638.1 | 18 | 2357 | |||
XM_008770417.1 | XP_008768639.1 | 17 | 2315 | |||
GenBank mRNA | HM801041.1 | - | 18 | 2243 | ||
M37394.2 | - | 18 | 2278 |