Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031561.2 | NP_113749.2 | 506 | |||
GenBank mRNA | AB005743.1 | - | 568 | |||
AF072411.1 | - | 2, 3 | 297 | |||
AF111268.1 | - | 313 | ||||
AF113914.1 | - | 2, 3 | 291 | |||
BC072543.1 | - | 506 | 7105943 | |||
L19658.1 | - | 2, 3 | 346 |
Gene Symbol: LOC100365047 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762678.1 | XP_008760900.1 | 2, 3 | 180 |
Gene Symbol: LOC103690020 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006235962.2 | XP_006236024.1 | 4, 5 | 932 | ||
XM_006235963.2 | XP_006236025.1 | 505 | ||||
XM_006235964.2 | XP_006236026.1 | 4, 5 | 948 | |||
XM_006235965.2 | XP_006236027.1 | 4, 5 | 937 |
Gene Symbol: RGD1565355 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109218.1 | NP_001102688.1 | 297 | |||
GenBank mRNA | BC091104.1 | - | 495 | 7383442 |