Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabggta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031654.2 | NP_113842.1 | 4 | 227 | ||
XM_006252009.2 | XP_006252071.1 | 4 | 322 | |||
XM_006252010.2 | XP_006252072.1 | 4 | 269 | |||
GenBank mRNA | BC086547.1 | - | 4 | 227 | 7301723 | |
L10415.1 | - | 187 | ||||
S62096.1 | - | 187 |