Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apba3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031781.1 | NP_113969.1 | 1 | 564 | ||
XM_006241018.2 | XP_006241080.1 | 2 | 725 | |||
XM_006241019.2 | XP_006241081.1 | 2 | 698 | |||
XM_006241020.2 | XP_006241082.1 | 2 | 723 | |||
GenBank mRNA | AF029109.1 | - | 1 | 564 | ||
BC161842.1 | - | 2 | 626 | 9102177 |