Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfe2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031789.2 | NP_113977.1 | 2 | 279 | ||
XM_006234396.2 | XP_006234458.1 | 2 | 427 | |||
XM_006234397.2 | XP_006234459.1 | 2 | 372 | |||
XM_006234398.2 | XP_006234460.1 | 2 | 979 | |||
GenBank mRNA | AF037350.1 | - | 2 | 281 | ||
BC061724.1 | - | 2 | 294 | 5598313 | ||
FQ221441.1 | - | 2 | 300 |