Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Omd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031817.1 | NP_114005.1 | 1 | 197 | ||
XM_008771551.1 | XP_008769773.1 | 1 | 1092 | |||
GenBank mRNA | AF104362.1 | - | 1 | 197 |
Gene Symbol: LOC103690116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771520.1 | XP_008769742.1 | 3014 | |||
XM_008771521.1 | XP_008769743.1 | 1092 |