Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031841.1 | NP_114029.1 | 6 | 4822 | ||
GenBank mRNA | AB032243.1 | - | 6 | 4822 | ||
BC061737.1 | - | 6 | 4757 | 5599134 | ||
FQ212986.1 | - | 1 | 906 | |||
FQ213614.1 | - | 1 | 693 |