Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nae1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC127481.1 | - | 18 | 1479 | 8370668 | |
FQ234218.1 | - | 7 | 574 | |||
U90829.1 | - | 18 | 1476 |