Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F2rl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053313.1 | NP_445765.1 | 2 | 1014 | ||
GenBank mRNA | AF310076.1 | - | 2 | 1014 |