Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053425.1 | NP_445877.1 | 3, 4 | 298 | ||
GenBank mRNA | AF255305.1 | - | 3, 4 | 298 | ||
BC158586.1 | - | 3, 4 | 258 | 9099085 |