Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fen1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053430.1 | NP_445882.1 | 2 | 2101 | ||
XM_006231072.2 | XP_006231134.1 | 3 | 2072 | |||
XM_006231073.2 | XP_006231135.1 | 2 | 2096 | |||
XM_006231074.2 | XP_006231136.1 | 2 | 1903 | |||
XM_008760245.1 | XP_008758467.1 | 2 | 1886 | |||
XM_008760246.1 | XP_008758468.1 | 2 | 1886 | |||
GenBank mRNA | AF281018.1 | - | 2 | 2101 | ||
BC083630.1 | - | 2 | 2073 | 7134402 |