Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xrcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053435.1 | NP_445887.1 | 10 | 1217 | ||
XM_006228481.2 | XP_006228543.2 | 15 | 1856 | |||
XM_008758991.1 | XP_008757213.1 | 13 | 1718 | |||
GenBank mRNA | AF290895.1 | - | 10 | 1217 | ||
BC070894.1 | - | 10 | 1194 | 7099372 |