Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053442.1 | NP_445894.1 | 10 | 2028 | ||
GenBank mRNA | AB024400.1 | - | 10 | 2028 |