Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053457.2 | NP_445909.2 | 3 | 767 | ||
GenBank mRNA | AF324043.1 | - | 3 | 572 | ||
BC070927.1 | - | 3 | 767 | 7096642 |