Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kpna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270802.1 | NP_001257731.1 | 3 | 290 | ||
NM_053483.2 | NP_445935.2 | 3 | 265 | |||
GenBank mRNA | AJ130946.1 | - | 2 | 169 | ||
AY779026.1 | - | 2 | 159 | |||
BC089787.1 | - | 3 | 216 | 7326072 | ||
FM083931.1 | - | 3 | 265 | |||
FM115539.1 | - | 3 | 287 |