Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gas7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053484.2 | NP_445936.2 | 2, 3 | 136 | ||
XM_006246827.2 | XP_006246889.1 | 2, 3 | 805 | |||
XM_006246828.2 | XP_006246890.1 | 2, 3 | 1646 | |||
XM_006246829.2 | XP_006246891.1 | 2, 3 | 989 | |||
XM_006246830.2 | XP_006246892.1 | 2, 3 | 320 | |||
XM_008767883.1 | XP_008766105.1 | 2, 3 | 720 | |||
GenBank mRNA | AJ003148.1 | - | 2, 3 | 272 | ||
AJ131902.1 | - | 272 |