Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOPBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007027.3 | NP_008958.2 | 5 | 585 | ||
XM_005247076.3 | XP_005247133.1 | 5 | 1072 | |||
XM_011512357.2 | XP_011510659.1 | 5 | 656 | |||
XM_017005636.1 | XP_016861125.1 | 5 | 655 | |||
XM_017005637.1 | XP_016861126.1 | 2 | 191 | |||
GenBank mRNA | AB019397.1 | - | 5 | 532 | ||
AK302584.1 | - | 5 | 551 | |||
BC032633.1 | - | 5 | 596 | |||
BC126209.1 | - | 4 | 475 | 8991925 | ||
BC151237.1 | - | 5 | 538 | 8860504 | ||
D87448.1 | - | 5 | 538 |