Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STMN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199214.1 | NP_001186143.1 | 3 | 546 | ||
NM_007029.3 | NP_008960.2 | 3 | 546 | |||
XM_005251142.2 | XP_005251199.1 | 4 | 170 | |||
GenBank mRNA | AK292737.1 | - | 3 | 218 | ||
AK297485.1 | - | 3 | 227 | |||
AK297594.1 | - | 3 | 240 | |||
AK299500.1 | - | 3 | 255 | |||
AK308557.1 | - | 3 | 229 | |||
AK309130.1 | - | 3 | 433 | |||
BC006302.2 | - | 3 | 215 | 4100671 | ||
BQ069488.1 | - | 3 | 208 | |||
BT020034.1 | - | 2 | 164 | |||
CR456833.1 | - | 2 | 164 | |||
D50375.1 | - | 3 | 281 | |||
S82024.1 | - | 3 | 192 |